Thermo Scientific Chemicals

Tetraammineplatinum(II) nitrate, Thermo Scientific Chemicals

Catalog number: 088960.03
1 g, Each
Thermo Scientific Chemicals

Tetraammineplatinum(II) nitrate, Thermo Scientific Chemicals

Catalog number: 088960.03
1 g, Each
Quantity
Catalog number: 088960.03
also known as 088960-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20634-12-2
IUPAC Name
platinum(2+) tetraamine dinitrate
Molecular Formula
H12N6O6Pt
InChI Key
RBAKORNXYLGSJB-UHFFFAOYSA-N
SMILES
N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
White
Form
Powder/chunks
Assay (special)
Pt 49.9-50.7%

Description

Tetraammineplatinum(II) nitrate is widely utilized as a precursor to bimetallic Pt/Zn catalysts for the selective hydrogenation of crotonaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text