1x1 image pixel for data collection
Dionex™ Retention Time Standards for Ion Chromatography
Dionex™ Retention Time Standards for Ion Chromatography
Actual product may vary
Dionex™ Retention Time Standards for Ion Chromatography
Dionex™ Retention Time Standards for Ion Chromatography
Thermo Scientific™
Dionex™ Retention Time Standards for Ion Chromatography
Automate the task of entering and updating component retention times using Thermo Scientific™ Dionex™ Retention Time Standards, available for anion and cation analyses.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberStandard ConcentrationFor Use With (Application)Standard Components
088964LowRoutine Cation DeterminationsLithium, Sodium, Ammonium, Potassium, Magnesium, Calcium
088957LowRoutine Anion DeterminationsFluoride, Chloride, Nitrite, Bromide, Nitrate, Phosphate
302511HighRoutine Anion DeterminationsFluoride, Chloride, Nitrite, Bromide, Nitrate, Phosphate
088969HighRoutine Cation DeterminationsLithium, Sodium, Ammonium, Potassium, Magnesium, Calcium
Catalog number 088964
Price (USD)
214.00
Each
Add to cart
Standard Concentration:
Low
For Use With (Application):
Routine Cation Determinations
Standard Components:
Lithium, Sodium, Ammonium, Potassium, Magnesium, Calcium
Price (USD)
214.00
Each
Add to cart
Automate the task of entering and updating component retention times and names using Thermo Scientific™ Dionex™ Retention Time Standards for ion chromatography. Available for both anion and cation analyses, these standards contain specific ratios of analytes and have been developed for use with Thermo Scientific™ Dionex™ Chromeleon™ 7.2 software retention time standard (RTS) processing methods.
How do they work?
Predefined components with assigned retention times are interpreted as peak area ratios. The peak area ratio for a component is its peak area percentage relative to the largest peak in the retention time standard. For example, if the largest peak has an area of 0.09, and peak 5 has an area of 0.045, then the ratio of peak 5 to the reference (largest) peak is 50%. Peak area ratios allow dynamic determination of expected component retention times. Even with a shift in retention time, Chromeleon software automatically recognizes and correctly assigns components based on these peak area ratios.
When a sequence is run, the retention time standard is included in the run and the injection is identified as a retention time standard. When the sequence data is processed, Chromeleon verifies that the retention time standard is valid based on predefined criteria. For each injection in a sequence that is assigned an RTS processing method, Chromeleon searches for the first valid retention time standard with the same assigned processing method. This is designated as the reference retention time standard for the injection. The retention times of the identified components from the reference retention time standard are then used as the expected retention times to identify components in unknown injections that reference this retention time standard.
Increase Your Productivity
  • Dynamic determination of expected retention times based on peak area ratios.
  • Easy and convenient IC standard preparation.
  • Reduce standard preparation time.
  • Reduce time-consuming standard validation.
  • Reduce time required to compensate for factors like variations in run conditions.
  • Easy access to default method templates that contain predefined component names and peak ratios using Chromeleon 7.2 SR3 or later.
Guarantee the Reliability of Your Analyses
  • Reduce standard preparation errors.
  • Increase accuracy of quantifications.
  • Improve day-to-day reproducibility.
The Quality You Expect from Thermo Fisher Scientific
  • Uses only high-purity, reagent-grade chemicals and water (18MΩ-cm), quality-control checked for ionic contamination.
  • All standards are National Institute of Standards and Technology (NIST) traceable.
  • A Certificate of Analysis is provided with every standard.
Unconditional Guarantee
Your satisfaction is guaranteed. All Thermo Scientific Dionex standards carry a 90-day warranty.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Application)Routine Cation Determinations
QuantityEach
Standard ComponentsLithium, Sodium, Ammonium, Potassium, Magnesium, Calcium
Standard ConcentrationLow
StorageRoom Temperature
Volume (Metric)50 mL
TypeStandard
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link