Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, iron(III) monosodium salt, Thermo Scientific Chemicals

Catalog number: 088995.A3
2 kg, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, iron(III) monosodium salt, Thermo Scientific Chemicals

Catalog number: 088995.A3
2 kg, Each
Quantity
Catalog number: 088995.A3
also known as 088995-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15708-41-5
IUPAC Name
iron(3+) sodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Molecular Formula
C10H12FeN2NaO8
InChI Key
MKWYFZFMAMBPQK-UHFFFAOYSA-J
SMILES
[Na+].[Fe+3].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Assay (Titration)
Total EDTA: ≥65.5%
Form
powder
Appearance (Color)
Pale yellow
Assay (Titration)
Fe (III) 12.5 - 13.5%

Description

Ethylenediaminetetraacetic acid, iron(III) monosodium salt is used to eliminate enzyme inhibition by traces of heavy metals and to inhibit enzymes that require divalent cations as cofactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text