Magnesium hexafluorosilicate hydrate, Thermo Scientific Chemicals
Magnesium hexafluorosilicate hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium hexafluorosilicate hydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 089018.36
also known as 089018-36
Price (USD)/ Each
68.60
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
68.60
Add to cart
Magnesium hexafluorosilicate hydrate, Thermo Scientific Chemicals
Catalog number089018.36
Price (USD)/ Each
68.60
-
Add to cart
Specifications
Chemical Name or MaterialMagnesium hexafluorosilicate hydrate
Melting Point120°C (decomposition)
Health Hazard 1H301
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3P264b-P270-P301+P310-P330-P501c
View more
It is employed as an antiplaque agents and oral care agents. Also used for mothproofing textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only