Thermo Scientific Chemicals

Nickel(II) sulfamate hydrate, Thermo Scientific Chemicals

Catalog number: 089020.30
250 g, Each
Thermo Scientific Chemicals

Nickel(II) sulfamate hydrate, Thermo Scientific Chemicals

Catalog number: 089020.30
250 g, Each
Quantity
Catalog number: 089020.30
also known as 089020-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13770-89-3
IUPAC Name
nickel(2+) disulfamate
Molecular Formula
H4N2NiO6S2
InChI Key
KERTUBUCQCSNJU-UHFFFAOYSA-L
SMILES
[Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O
Assay (unspecified)
15.1%-19.2% Ni

Description

Nickel(II) sulfamate is used as an electro plating and surface treating agent. It finds application in metal coloring and casting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text