Thermo Scientific Chemicals

Potassium hexafluorosilicate, 98%, Thermo Scientific Chemicals

Catalog number: 089023.A7
5 kg, Each
Thermo Scientific Chemicals

Potassium hexafluorosilicate, 98%, Thermo Scientific Chemicals

Catalog number: 089023.A7
5 kg, Each
Quantity
Catalog number: 089023.A7
also known as 089023-A7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16871-90-2
IUPAC Name
dipotassium hexafluorosilanediuide
Molecular Formula
F6K2Si
InChI Key
UUHPPUWEDCVPCO-UHFFFAOYSA-N
SMILES
[K+].[K+].F[Si--](F)(F)(F)(F)F
Assay (unspecified)
98.6% min

Description

Potassium hexafluorosilicate is used in the production of porceleins. It is also used for the preservation of timber. It is involved in the manufacturing of ceramics, aluminum and magnesium smelting. In addition, it is used as an intermediate in the manufacturing of optical glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text