Thermo Scientific Chemicals

Sodium trimetaphosphate, Thermo Scientific Chemicals

Catalog number: 089063.A1
1 kg, Each
Thermo Scientific Chemicals

Sodium trimetaphosphate, Thermo Scientific Chemicals

Catalog number: 089063.A1
1 kg, Each
Quantity
Catalog number: 089063.A1
also known as 089063-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7785-84-4
IUPAC Name
trisodium trioxo-1,3,5,2λ⁵,4λ⁵,6λ⁵-trioxatriphosphinane-2,4,6-tris(olate)
Molecular Formula
Na3O9P3
InChI Key
UGTZMIPZNRIWHX-UHFFFAOYSA-K
SMILES
[Na+].[Na+].[Na+].[O-]P1(=O)OP([O-])(=O)OP([O-])(=O)O1
Assay (unspecified)
P₂O₅ 68.0-70.0

Description

Sodium trimetaphosphate is used as a crosslinking agent in the cross-linked amylase-resistant starch. It acts as an intermediate in food industries. Further, it serves as a corrosion inhibitors, anti-scaling agents, fillers, finishing agents, plating agents and surface treating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text