Thermo Scientific Chemicals

Aluminum bromide, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 089065.14
25 g, Each
Thermo Scientific Chemicals

Aluminum bromide, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 089065.14
25 g, Each
Quantity
Catalog number: 089065.14
also known as 089065-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7727-15-3
IUPAC Name
aluminium(3+) tribromide
Molecular Formula
AlBr3
InChI Key
PQLAYKMGZDUDLQ-UHFFFAOYSA-K
SMILES
[Al+3].[Br-].[Br-].[Br-]
Total Metal Impurities
0.003% max.

Description

Aluminum Bromide, is used as a catalyst for the Friedel-Crafts alkylation reaction, and also for bromination and isomerization in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text