Thermo Scientific Chemicals

Amberlyst™ 15(H), ion exchange resin, Thermo Scientific Chemicals

Catalog number: 089079.A1
1 kg, Each
Thermo Scientific Chemicals

Amberlyst™ 15(H), ion exchange resin, Thermo Scientific Chemicals

Catalog number: 089079.A1
1 kg, Each
Quantity
Catalog number: 089079.A1
also known as 089079-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39389-20-3
IUPAC Name
1,2-diethenylbenzene; 2-ethenylbenzene-1-sulfonic acid
Molecular Formula
C18H18O3S
InChI Key
SIWVGXQOXWGJCI-UHFFFAOYSA-N
SMILES
C=CC1=CC=CC=C1C=C.OS(=O)(=O)C1=CC=CC=C1C=C
Exchange capacity
Total capacity >/= 4.70 eq/Kg
Residual water
< / = 1.6%

Description

Amberlyst™ 15(H) is a strongly acidic cation exchange resin used as a heterogeneous acid catalysis and is suitable for non-aqueous catalysis. It is used to determine the mechanism of phenol alkylation using density functional theory (DFT). It is widely utilized as a column packing material for HPLC analysis. It is employed in separation media for Thin-layer chromatography (TLC) and paper chromatography. It is used in various chemical reactions like esterification, transesterification, Michael addition, Prins cyclization, Friedel-Crafts alkylation, acylation, cleavage of epoxides and crossed-aldol condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text