Thermo Scientific Chemicals

Gadolinium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 089158.06
5 g, Each
Thermo Scientific Chemicals

Gadolinium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 089158.06
5 g, Each
Quantity
Catalog number: 089158.06
also known as 089158-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100587-93-7
IUPAC Name
gadolinium(3+) triacetate
Molecular Formula
C6H9GdO6
InChI Key
LYQGMALGKYWNIU-UHFFFAOYSA-K
SMILES
[Gd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Gadolinium(III) acetate hydrate is used as a laboratory reagent, catalysts, optical glasses, structural ceramics, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text