Search Thermo Fisher Scientific
Aluminum di(sec-butoxide)acetoacetic ester chelate is used in addition with lithium acetate and titanium(IV) ethoxide to prepare precursor sol to prepare Li1.3Al0.3Ti1.7(PO4)3 thin films on glass substrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.