Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min, Thermo Scientific Chemicals
Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 089350.22
also known as 089350-22
Price (USD)/ Each
70.40
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
70.40
Add to cart
Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min, Thermo Scientific Chemicals
Catalog number089350.22
Price (USD)/ Each
70.40
-
Add to cart
Chemical Identifiers
CAS14782-75-3
IUPAC Namealuminium(3+) 1-ethoxy-1,3-dioxobutan-2-ide bis(propan-2-olate)
Molecular FormulaC12H23AlO5
InChI KeyAHPLRRISNPYWTH-UHFFFAOYSA-N
SMILES[Al+3].CC(C)[O-].CC(C)[O-].CCOC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofAAl: 9.6-9.8%
CommentMay crystallize on storage.
Appearance (Color)Clear pale yellow
FormViscous liquid
Aluminum di(isopropoxide)acetoacetic ester chelate is used as a gel ink. It finds application in paint to give better drying, luster and hardness, and improve the performance as surface wrinkling. It is also used as a crosslinking agent in epoxy, alkyd and polyester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only