Thermo Scientific Chemicals

Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min, Thermo Scientific Chemicals

Catalog number: 089350.36
500 g, Each
Thermo Scientific Chemicals

Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min, Thermo Scientific Chemicals

Catalog number: 089350.36
500 g, Each
Quantity
Catalog number: 089350.36
also known as 089350-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14782-75-3
IUPAC Name
aluminium(3+) 1-ethoxy-1,3-dioxobutan-2-ide bis(propan-2-olate)
Molecular Formula
C12H23AlO5
InChI Key
AHPLRRISNPYWTH-UHFFFAOYSA-N
SMILES
[Al+3].CC(C)[O-].CC(C)[O-].CCOC(=O)[CH-]C(C)=O
Assay from Supplier's CofA
Al: 9.6-9.8%
Comment
May crystallize on storage.
Appearance (Color)
Clear pale yellow
Form
Viscous liquid

Description

Aluminum di(isopropoxide)acetoacetic ester chelate is used as a gel ink. It finds application in paint to give better drying, luster and hardness, and improve the performance as surface wrinkling. It is also used as a crosslinking agent in epoxy, alkyd and polyester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text