Silicon(IV) oxide, amorphous fumed, S.A. 350-420m2/g, Thermo Scientific Chemicals
Silicon(IV) oxide, amorphous fumed, S.A. 350-420m2/g, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicon(IV) oxide, amorphous fumed, S.A. 350-420m2/g, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 089376.18
also known as 089376-18
Price (USD)/ Each
19.65
Online exclusive
21.90 
Save 2.25 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
19.65
Online exclusive
21.90 
Save 2.25 (10%)
Add to cart
Silicon(IV) oxide, amorphous fumed, S.A. 350-420m2/g, Thermo Scientific Chemicals
Catalog number089376.18
Price (USD)/ Each
19.65
Online exclusive
21.90 
Save 2.25 (10%)
-
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Surface Area350-420m²/g
Functionalized RAFT agent for controlled radical polymerization; especially suited for the polymerization of styrene; acrylate and acrylamide monomers. Azide group can be used to conjugate to a variety of alkyne-functionalized biomolecules. Chain Transfer Agent (CTA). SDS mixture of sodium alkyl sulfates consisting chiefly of sodium lauryl sulfate. Used as a suspending and thickening agent in pharmaceutical

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only