Thermo Scientific Chemicals

Silicon(IV) oxide, amorphous fumed, S.A. 350-420m2/g, Thermo Scientific Chemicals

Catalog number: 089376.A1
1 kg, Each
Thermo Scientific Chemicals

Silicon(IV) oxide, amorphous fumed, S.A. 350-420m2/g, Thermo Scientific Chemicals

Catalog number: 089376.A1
1 kg, Each
Quantity
Catalog number: 089376.A1
also known as 089376-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Surface Area
350-420m²/g

Description

Functionalized RAFT agent for controlled radical polymerization; especially suited for the polymerization of styrene; acrylate and acrylamide monomers. Azide group can be used to conjugate to a variety of alkyne-functionalized biomolecules. Chain Transfer Agent (CTA). SDS mixture of sodium alkyl sulfates consisting chiefly of sodium lauryl sulfate. Used as a suspending and thickening agent in pharmaceutical

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text