Thermo Scientific Chemicals

Phosphorous acid, 97%, Thermo Scientific Chemicals

Catalog number: 089407.36
500 g, Each
Thermo Scientific Chemicals

Phosphorous acid, 97%, Thermo Scientific Chemicals

Catalog number: 089407.36
500 g, Each
Quantity
Catalog number: 089407.36
also known as 089407-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13598-36-2
IUPAC Name
dihydroxy(oxo)-λ⁵-phosphanylium
Molecular Formula
H2O3P
InChI Key
XNQULTQRGBXLIA-UHFFFAOYSA-O
SMILES
O[P+](O)=O
Assay (unspecified)
96.1% min

Description

Phosphorus acid is used as an intermediate in the preparation of other phosphorus compounds like potassium phosphite, ammonium phosphite and calcium phosphite. It is actively involved in the preparation of phosphites like aminotris(methylenephosphonic acid) (ATMP), 1-hydroxyethane 1,1-diphosphonic acid (HEDP) and 2-phosphonobutane-1,2,4-tricarboxylic Acid (PBTC), which find application in water treatment as a scale or corrosive inhibitor. It is also used in chemical reactions as a reducing agent. Its salt, lead phosphite is used as PVC stabilizer. It is also used as a precursor in the preparation of phosphine and as an intermediate in the preparation of other phosphorus compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text