Thermo Scientific Chemicals

Sodium isopropoxide, Thermo Scientific Chemicals

Catalog number: 089446.06
5 g, Each
Thermo Scientific Chemicals

Sodium isopropoxide, Thermo Scientific Chemicals

Catalog number: 089446.06
5 g, Each
Quantity
Catalog number: 089446.06
also known as 089446-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
683-60-3
IUPAC Name
sodium propan-2-olate
Molecular Formula
C3H7NaO
InChI Key
WBQTXTBONIWRGK-UHFFFAOYSA-N
SMILES
[Na+].CC(C)[O-]
Appearance (Color)
White to pale cream to pale yellow
Form
Powder
Chloride content
≤0.01%
Total Metal Impurities
≤1 %

Description

Sodium isopropoxide is used in the preparation of ibuprofen intermediate. It is involved in reactions such as polymerization of butadiene. Further, it acts as a catalyst in the manufacturing of acetone and its derivatives. In addition, it is used as a solvent and serves as an antiseptic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text