Thermo Scientific Chemicals

Titanium(IV) isobutoxide, Thermo Scientific Chemicals

Catalog number: 089455.18
50 g, Each
Thermo Scientific Chemicals

Titanium(IV) isobutoxide, Thermo Scientific Chemicals

Catalog number: 089455.18
50 g, Each
Quantity
Catalog number: 089455.18
also known as 089455-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7425-80-1
IUPAC Name
titanium(4+) tetrakis(2-methylpropan-1-olate)
Molecular Formula
C16H36O4Ti
InChI Key
QUVMSYUGOKEMPX-UHFFFAOYSA-N
SMILES
[Ti+4].CC(C)C[O-].CC(C)C[O-].CC(C)C[O-].CC(C)C[O-]
Form
Liquid to low melting solid
Infrared spectrum
Conforms
Appearance (Color)
Pale yellow

Description

Titanium(IV) isobutoxide general application in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text