Chloroform-d, 99.8% (Isotopic), Thermo Scientific Chemicals
Chloroform-d, 99.8% (Isotopic), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloroform-d, 99.8% (Isotopic), Thermo Scientific Chemicals

 
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 089541.18
also known as 089541-18
Price (USD)/ Each
36.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
43.40 
Save 6.75 (16%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
36.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
43.40 
Save 6.75 (16%)
Add to cart
Chloroform-d, 99.8% (Isotopic), Thermo Scientific Chemicals
Catalog number089541.18
Price (USD)/ Each
36.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
43.40 
Save 6.75 (16%)
-
Add to cart
Chemical Identifiers
CAS865-49-6
IUPAC Nametrichloro(²H)methane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-MICDWDOJSA-N
SMILES[2H]C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Proton NMRIsotopic enrichment >/= 99.8%
Chloroform-d is widely used in the organic solvent for the NMR analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only