Thermo Scientific Chemicals

Potassium bis(oxalato)oxotitanate(IV) dihydrate, Thermo Scientific Chemicals

Catalog number: 089620.36
500 g, Each
Thermo Scientific Chemicals

Potassium bis(oxalato)oxotitanate(IV) dihydrate, Thermo Scientific Chemicals

Catalog number: 089620.36
500 g, Each
Quantity
Catalog number: 089620.36
also known as 089620-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14402-67-6
IUPAC Name
dipotassium oxotitaniumbis(ylium) dioxalate
Molecular Formula
C4K2O9Ti
InChI Key
UHWHMHPXHWHWPX-UHFFFAOYSA-J
SMILES
[K+].[K+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
Assay (unspecified)
>/= 90%

Description

Mordant in dyeing, reagent Potassium bis(oxalato)oxotitanate(IV) dihydrate is used as a mordant in dyeing and acts as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text