Thermo Scientific Chemicals

Lithium aluminum hydride, pellets, 97%, Thermo Scientific Chemicals

Catalog number: 089643.22
100 g, Each
Thermo Scientific Chemicals

Lithium aluminum hydride, pellets, 97%, Thermo Scientific Chemicals

Catalog number: 089643.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 089643.22
also known as 089643-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16853-85-3
IUPAC Name
lithium(1+) alumanuide
Molecular Formula
AlH4Li
InChI Key
OCZDCIYGECBNKL-UHFFFAOYSA-N
SMILES
[Li+].[AlH4-]
Assay (unspecified)
96.1% min

Description

Lithium aluminum hydride is used as a reducing agent in various synthetic organic chemistry reactions. For example, it is used in the conversion of esters, carboxylic acids, acyl chlorides, aldehydes and ketones into their corresponding alcohols. It also involved in the conversion of amide, nitro, nitrile, imine, oxime and azide compounds into their amines as well as reduces alkyl halides to alkanes. It reacts with sodium chloride to prepare sodium hydride. It acts as a potential hydrogen storage medium which could be used for future fuel cell-powered vehicles. It is mainly used in the preparation of other tetrahydridoaluminumates like sodium aluminum hydride, potassium aluminum hydride and magnesium alanate. Furthermore, it has been used to prepare thermoplastic polyester polyamides from oleic acid and lithium-polymer batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text