Thermo Scientific Chemicals

Tin(II) tetrafluoroborate, 50% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 089669.D5
3 x 1 kg, Each
Thermo Scientific Chemicals

Tin(II) tetrafluoroborate, 50% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 089669.D5
3 x 1 kg, Each
Quantity
Catalog number: 089669.D5
also known as 089669-D5
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13814-97-6
IUPAC Name
λ²-tin(2+) bis(tetrafluoroboranuide)
Molecular Formula
B2F8Sn
InChI Key
JPZSNSDMGTYJIW-UHFFFAOYSA-N
SMILES
[Sn++].F[B-](F)(F)F.F[B-](F)(F)F
Form
Liquid (aqueous solution)
Assay (unspecified)
49.0-52.0%

Description

Tin(II) tetrafluoroborate is used in electroplating. It is also used for plating bath tinned copper, tin and tin alloy. Further, it acts as a catalyst for various organic reactions. It is also employed in metal plating. In addition to this, it is used as a petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text