Thermo Scientific Chemicals

Niobium(IV) oxide, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 089692.09
10 g, Each
Thermo Scientific Chemicals

Niobium(IV) oxide, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 089692.09
10 g, Each
Quantity
Catalog number: 089692.09
also known as 089692-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12034-59-2
Appearance (Color)
Dark grey to black
Form
Powder
Total Metal Impurities
≤1.0% (≤10000ppm)

Description

Niobium(IV) oxide is used as a reducing agent, which reduces carbon dioxide and sulfur dioxide to carbon and sulfur respectively. It finds application in dielectric material for electronic capacitors. It is also used for manufacturing metal niobium metal, in optical glass and in electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text