Thermo Scientific™

Drierite™, indicating (CaSO{4}), Thermo Scientific Chemicals

Catalog number: 089751.P5
5 lb., Each
Thermo Scientific™

Drierite™, indicating (CaSO{4}), Thermo Scientific Chemicals

Catalog number: 089751.P5
5 lb., Each
Quantity
Catalog number: 089751.P5
also known as 089751-P5
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7778-18-9
IUPAC Name
calcium sulfate
Molecular Formula
CaO4S
InChI Key
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES
[Ca++].[O-]S([O-])(=O)=O
Comment
-8 Mesh
Appearance (Color)
Blue
Form
Granules
Assay from Supplier's CofA
Moisture: ≤0.5%

Description

Drierite™, indicating (CaSO{4}) is used in drying of air, industrial gases, refrigerants and many organic liquids. It is used to maintain a dry atmosphere in storage spaces, vaults and commercial packages. It is utilized for the protection of hygroscopic materials, corrosion, rust, and other deterioration caused by high humidity. Further, it is used as an additive in polymer formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text