Tin(II) pyrophosphate, Thermo Scientific Chemicals
Tin(II) pyrophosphate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(II) pyrophosphate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 089785.18
also known as 089785-18
Price (USD)/ Each
48.80
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
48.80
Add to cart
Tin(II) pyrophosphate, Thermo Scientific Chemicals
Catalog number089785.18
Price (USD)/ Each
48.80
-
Add to cart
Chemical Identifiers
CAS15578-26-4
IUPAC Namebis(λ²-tin(2+)) (phosphonatooxy)phosphonate
Molecular FormulaO7P2Sn2
InChI KeyGEZAUFNYMZVOFV-UHFFFAOYSA-J
SMILES[Sn++].[Sn++].[O-]P([O-])(=O)OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96% min
Tin(II) pyrophosphate is used to inhibit the formation of volatile sulfur compounds. Further, it is used for cyanide-free tin coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only