Thermo Scientific Chemicals

Vanadium(V) triisopropoxide oxide, 96%, Thermo Scientific Chemicals

Catalog number: 089798.06
5 g, Each
Thermo Scientific Chemicals

Vanadium(V) triisopropoxide oxide, 96%, Thermo Scientific Chemicals

Catalog number: 089798.06
5 g, Each
Quantity
Catalog number: 089798.06
also known as 089798-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5588-84-1
Molecular Formula
C9H24O4V
InChI Key
JOUSPCDMLWUHSO-UHFFFAOYSA-N
SMILES
O=[V].CC(C)O.CC(C)O.CC(C)O
Molecular Weight (g/mol)
247.23
Appearance (Color)
Clear yellow to orange to pale brown or green to gold
Assay (Titration)
≥95.0% (Vanadium-51 NMR, non-U.S. specification)
Assay from Suppliers CofA
≥95.0% (U.S. specification)
Comment
May darken on storage
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

Vanadium(V) triisopropoxide oxide serves as a building block for a series of neutral oxovanadium complexes containing both ethoxide and acetylacetonate ligands. It is also used in catalysis. Further, it acts as a precursor and used in thermal atomic layer deposition of vanadium pentoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text