Lead(II) 2,4-pentanedionate, Thermo Scientific Chemicals
Lead(II) 2,4-pentanedionate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) 2,4-pentanedionate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 089858.09
also known as 089858-09
Price (USD)/ Each
100.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
118.00 
Save 17.35 (15%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
100.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
118.00 
Save 17.35 (15%)
Add to cart
Lead(II) 2,4-pentanedionate, Thermo Scientific Chemicals
Catalog number089858.09
Price (USD)/ Each
100.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
118.00 
Save 17.35 (15%)
-
Add to cart
Chemical Identifiers
CAS15282-88-9
IUPAC Nameλ²-lead(2+) bis(2,4-dioxopentan-3-ide)
Molecular FormulaC10H14O4Pb
InChI KeyVJRBSSRKZYRKTG-UHFFFAOYSA-N
SMILES[Pb++].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to yellow to brown
FormSolid
Identification (FTIR)Conforms
Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only