Lutetium, AAS standard solution, Specpure™ Lu 1000μg/mL, Thermo Scientific Chemicals
Lutetium, AAS standard solution, Specpure™ Lu 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lutetium, AAS standard solution, Specpure™ Lu 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number 089886.AP
also known as 089886-AP
Price (USD)/ Each
224.00
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
224.00
Add to cart
Lutetium, AAS standard solution, Specpure™ Lu 1000μg/mL, Thermo Scientific Chemicals
Catalog number089886.AP
Price (USD)/ Each
224.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialLutetium
CAS12032-20-1
Health Hazard 1H290-H314-H335
Quantity500 mL
CAS7697-37-2
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only