Samarium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
Samarium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Samarium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 089916.09
also known as 089916-09
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
Add to cart
Samarium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
Catalog number089916.09
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10361-82-7
IUPAC Namesamarium(3+) trichloride
Molecular FormulaCl3Sm
InChI KeyBHXBZLPMVFUQBQ-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Sm+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Samarium(III) chloride is used in laboratories, preparation of samarium metals. The anhydrous chloride is used to prepare organometallic compounds of samarium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only