Erbium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Erbium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Erbium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 089917.09
also known as 089917-09
Price (USD)/ Each
78.65
Online exclusive
86.70 
Save 8.05 (9%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
78.65
Online exclusive
86.70 
Save 8.05 (9%)
Add to cart
Erbium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number089917.09
Price (USD)/ Each
78.65
Online exclusive
86.70 
Save 8.05 (9%)
-
Add to cart
Chemical Identifiers
CAS10138-41-7
IUPAC Nameerbium(3+) trichloride
Molecular FormulaCl3Er
InChI KeyHDGGAKOVUDZYES-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Er+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Total Metal Impurities≤0.1%
It is a starting material for new edge-bridged octahedral M6 cluster compounds which are used for electron and stability studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only