Thermo Scientific Chemicals

Tin(IV) acetate, Thermo Scientific Chemicals

Catalog number: 089921.06
5 g, Each
Thermo Scientific Chemicals

Tin(IV) acetate, Thermo Scientific Chemicals

Catalog number: 089921.06
5 g, Each
Quantity
Catalog number: 089921.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2800-96-6
IUPAC Name
tin(4+) tetraacetate
Molecular Formula
C8H12O8Sn
InChI Key
YJGJRYWNNHUESM-UHFFFAOYSA-J
SMILES
[Sn+4].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
Form
Powder / Crystalline powder
Elemental Analysis
%C=26.6 -27.0 ; %H=3.3 -3.5
Appearance (Color)
White
Gravimetric analysis
≥98.5% (%Sn=32.9 -34.0 after ignition to oxide)

Description

Tin(IV) acetate is used to prepare tin-based oxides (Sn-Zn-P-O), which is a negative electrodes for rechargeable lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text