Tin(IV) acetate, Thermo Scientific Chemicals
Tin(IV) acetate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(IV) acetate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 089921.14
also known as 089921-14
Price (USD)/ Each
701.65
Online exclusive
779.00 
Save 77.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
701.65
Online exclusive
779.00 
Save 77.35 (10%)
Add to cart
Tin(IV) acetate, Thermo Scientific Chemicals
Catalog number089921.14
Price (USD)/ Each
701.65
Online exclusive
779.00 
Save 77.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2800-96-6
IUPAC Nametin(4+) tetraacetate
Molecular FormulaC8H12O8Sn
InChI KeyYJGJRYWNNHUESM-UHFFFAOYSA-J
SMILES[Sn+4].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder / Crystalline powder
Elemental Analysis%C=26.6 -27.0 ; %H=3.3 -3.5
Appearance (Color)White
Gravimetric analysis≥98.5% (%Sn=32.9 -34.0 after ignition to oxide)
Tin(IV) acetate is used to prepare tin-based oxides (Sn-Zn-P-O), which is a negative electrodes for rechargeable lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only