Zinc molybdenum oxide, 98%, Thermo Scientific Chemicals
Zinc molybdenum oxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc molybdenum oxide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 089937.06
also known as 089937-06
Price (USD)/ Each
63.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
63.20
Add to cart
Zinc molybdenum oxide, 98%, Thermo Scientific Chemicals
Catalog number089937.06
Price (USD)/ Each
63.20
-
Add to cart
Chemical Identifiers
CAS13767-32-3
IUPAC Namezinc(2+) molybdenum tetraoxidandiide
Molecular FormulaMoO4Zn
InChI KeyCEKVBJSJDXQDQR-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[Zn++].[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.5% min
Used as a corrosion inhibitor in paints, adhesives, primer on metal surfaces, such as steel and aluminum. It may be applied by brush or spray, and is suitable as an after-blast or after-pickling, primer for plating, as an undercoat for alkyd enamels. Also used for dielectric and insulation applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only