Thermo Scientific Chemicals

Nickel molybdenum oxide, 98%, Thermo Scientific Chemicals

Catalog number: 089938.14
25 g, Each
Thermo Scientific Chemicals

Nickel molybdenum oxide, 98%, Thermo Scientific Chemicals

Catalog number: 089938.14
25 g, Each
Quantity
Catalog number: 089938.14
also known as 089938-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14177-55-0
IUPAC Name
nickel(2+) molybdenumbis(ylium)tetrakis(olate)
Molecular Formula
MoNiO4
InChI Key
AXSAYZUZQSGYFU-UHFFFAOYSA-N
SMILES
[Ni++].[O-][Mo++]([O-])([O-])[O-]
Assay (unspecified)
97.6% min

Description

Nickel molybdenum oxide is used as catalyst in petroleum industry for the oil fraction like naptha, kerosene and diesel. It is also used extensively as a chemical intermediate for catalytic purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text