Thermo Scientific Chemicals

Tungsten oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 089949.30
250 g, Each
Thermo Scientific Chemicals

Tungsten oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 089949.30
250 g, Each
Quantity
Catalog number: 089949.30
also known as 089949-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12037-58-0
Total Metal Impurities
0.01% max.

Description

Tungsten oxide is used as a pigment in ceramics and paints. It is used in various types of infrared absorbing coatings and foils. It is involved in the production of electrochromic windows, which is an electrically switchable glass that change light transmission properties with an applied voltage. It is a unique material utilized for various applications due their chromism, photocatalysis, and sensing capabilities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text