Thermo Scientific Chemicals

Tungsten dichloride dioxide, 99%, Thermo Scientific Chemicals

Catalog number: 089962.09
10 g, Each
Thermo Scientific Chemicals

Tungsten dichloride dioxide, 99%, Thermo Scientific Chemicals

Catalog number: 089962.09
10 g, Each
Quantity
Catalog number: 089962.09
also known as 089962-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13520-76-8
IUPAC Name
tungsten dichloride dioxidandiide
Molecular Formula
Cl2O2W
InChI Key
DVBXMVWZPVBSJY-UHFFFAOYSA-L
SMILES
[O--].[O--].[Cl-].[Cl-].[W]
Assay (unspecified)
98.6% min

Description

Tungsten dichloride dioxide is used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text