See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Orion™ BNC Meter Shorting Strap
Thermo Scientific™
Orion™ BNC Meter Shorting Strap
Use the Thermo Scientific™ Orion™ BNC Meter Shorting Strap on pH meters with BNC input to perform BNC input verification testing and meter self test
Have Questions?
Catalog number
090045
Price (USD)
/ Each
66.01
Special Offer
Online exclusive
Ends:
31-Dec-2024
77.20
Save 11.19 (14%)
-
Add to cart
Description:
Orion BNC shorting cap for pH, ORP and ISE meters
Price (USD)
/ Each
66.01
Special Offer
Online exclusive
Ends:
31-Dec-2024
77.20
Save 11.19 (14%)
Add to cart
Orion™ BNC Meter Shorting Strap
Catalog number
090045
Price (USD)
/ Each
66.01
Special Offer
Online exclusive
Ends:
31-Dec-2024
77.20
Save 11.19 (14%)
-
Add to cart
Use the Thermo Scientific™ Orion™ BNC Meter Shorting Strap on pH meters with BNC input to perform BNC input verification testing and meter self test.
Specifications
Description
Orion BNC shorting cap for pH, ORP and ISE meters
Type
Meter Accessories
Includes
One BNC shorting cap