Trimethyl(methylcyclopentadienyl)platinum(IV), Thermo Scientific Chemicals
Trimethyl(methylcyclopentadienyl)platinum(IV), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethyl(methylcyclopentadienyl)platinum(IV), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
0.5 g
Catalog number 098158.02
also known as 098158-02
Price (USD)/ Each
360.65
Online exclusive
401.00 
Save 40.35 (10%)
-
Add to cart
Quantity:
0.5 g
Request bulk or custom format
Price (USD)/ Each
360.65
Online exclusive
401.00 
Save 40.35 (10%)
Add to cart
Trimethyl(methylcyclopentadienyl)platinum(IV), Thermo Scientific Chemicals
Catalog number098158.02
Price (USD)/ Each
360.65
Online exclusive
401.00 
Save 40.35 (10%)
-
Add to cart
Chemical Identifiers
CAS94442-22-5
Molecular FormulaC9H16Pt
SMILESC[Pt](C)C.Cc1cccc1
Molecular Weight (g/mol)319.31
MDL NumberMFCD00079665
SpecificationsSpecification SheetSpecification Sheet
Assay by NMR≥95.0%
FormPowder
Appearance (Color)White to pale cream
Trimethyl(methylcyclopentadienyl)platinum(IV) is used as an organometallic precursor for platinum. It is used in the microelectronics, catalysis, photonics and chemical sensing processes. It is used for the deposition of ultrathin metallic layers of platinum by the electron beam induced deposition process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only