Search Thermo Fisher Scientific
Catalog Number | Description | Unit Size |
---|---|---|
1000-0010 | Imhoff Settling Cone | Case of 4 |
1000-0010PK also known as 1000-0010 | Imhoff Settling Cone | Each |
Determine settleable matter in water and waste effluent and also measure the sand content in well water. The Thermo Scientific™ Nalgene™ Polycarbonate Imhoff Settling Cone is constructed from durable polycarbonate. The cone features a polypropylene screw closure. The provided leakproof† closure makes it possible to withdraw sediment for gravimetric anaylsis, and also aids in cleaning. Closure design enables user to read graduations to zero.
Settling Cone Graduation Intervals
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Yes, we do offer replacements. Please contact Thermo Scientific Laboratory Products Essentials technical support for more information.
Share catalog number, name or link