Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
100022591 | 1 Syringe |
This 1 mL sample syringe is used with the fluidics system of the Attune™ NxT Acoustic Focusing Cytometer to move sample to the flow cell. Replacement of this syringe is recommended as part of instrument maintenance.
Share catalog number, name or link