Attune™ NxT Drip Tray
Product Image
Invitrogen™

Attune™ NxT Drip Tray

The Attune NxT Drip Tray is an optional accessory for the Attune NxT Acoustic Focusing Cytometer that helps prevent sampleRead more
Have Questions?
Catalog number 100023101
Price (USD)/ 1 tray
1,956.00
-
Add to cart
Price (USD)/ 1 tray
1,956.00
Add to cart
Attune™ NxT Drip Tray
Catalog number100023101
Price (USD)/ 1 tray
1,956.00
-
Add to cart
The Attune NxT Drip Tray is an optional accessory for the Attune NxT Acoustic Focusing Cytometer that helps prevent sample from spilling onto the laboratory bench if the sample tube becomes dislodged from the SIP tube. The tray is easily installed and removed with the use of magnets. The drip tray has an indented lip that will hold several milliliters of spilled sample.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAttune™
Quantity1 tray
Shipping ConditionRoom Temperature
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeDrip Tray
Unit Size1 tray
Contents & Storage
Store at room temperature.