Informational:Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Dynabeads™ Protein G for Immunoprecipitation

Catalog number: 10003D
Have Questions?
Invitrogen™

Dynabeads™ Protein G for Immunoprecipitation

Catalog number: 10003D
Have Questions?
Catalog Number
10003D
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
10003D1 mL
-
10004D5 mL
-
10009D50 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Dynabeads Protein G beads are uniform, 2.8-μm superparamagnetic beads with recombinant Protein G (∼17 kDa) covalently coupled to the surface. Dynabeads Protein G beads provide a great alternative to Sepharose resin or agarose resin for immunoprecipitation (IP) combined with your own antibody, and both manual and automated protocols are available.

Benefits of Dynabeads Protein G beads include:
Fast and easy—IP in less than 40 minutes with no columns, centrifugations, or time-consuming pre-clearing required
High performance—high target protein yield with low antibody consumption
Low background—very low non-specific binding with a high signal-to-noise ratio
High reproducibility—uniform beads ensure the most consistent results
Automation ready—automation protocols available with the KingFisher Sample Purification instruments

Gentle separation causes minimal physical stress to proteins
The magnetic separation technology utilized by Dynabeads Protein G is rapid and gentle, causing minimal physical stress to target proteins. This permits the isolation and concentration of labile composites that might otherwise dissociate or be damaged by proteases during long incubation times. Native protein conformation and large protein complexes are preserved.

Binding strength and antibody binding capacity
Dynabeads Protein G beads allow for the isolation of most mammalian immunoglobulins (Ig). The amount of Ig captured depends on the concentration of Ig in the starting sample and on the type and source of the Ig. 100 μL of Dynabeads Protein G will isolate approximately 25–30 μg human IgG from a sample containing 20–200 μg IgG/mL. Predominantly Fc-binding allows optimal Ig orientation.

Dynabeads magnetic beads are not porous in nature such as Sepharose or agarose resins. This means that all antibodies bind to the smooth outer surface of the beads and thus are available to bind the target protein. This translates to a low antibody consumption while obtaining a high yield of the target protein. The smooth bead surface is also responsible for the low non-specific binding that Dynabeads magnetic beads are known for.

Dynabeads immunoprecipitation protocol is fast and easy
The manual IP protocol is simple and can be performed in less than 40 minutes with 3 easy bind-incubate-elute steps. The procedure does not require any pre-clearing due to the low non-specific binding and uses very little antibody while still obtaining a high IP yield, thus saving both time and cost per IP.

The antibody-coated beads can be used in a variety of IP applications such as Co-IP, chromatin IP (ChIP/ChIP sequencing), RNA IP (RIP, RIP sequencing), small-scale IgG purification, and protein purification. The isolated pure target protein can be used in downstream western blot, mass spectrometry analysis, sequencing, etc.

Automation-ready for use with KingFisher purification systems
The 2.8-μm magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher sample purification systems or liquid handlers.

Commercial supply
Our manufacturing sites are ISO 13485-certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them the ideal choice for commercial supply. If you are in the process of customizing Dynabeads Protein G beads in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or see our Dynabeads OEM page.

Learn more about Dynabeads magnetic beads and KingFisher Sample Purification Systems:
• Dynabeads Protein G beads are also available as a 'ready-to-go' kit with buffers included
See our immunoprecipitation selection table, data, and references
Dynabeads magnetic beads selection guide
See magnets for Dynabeads magnetic beads separations
KingFisher automation protocols
Watch a video about the KingFisher Flex instrument

*Sepharose is a trademark of GE Healthcare Bio-Sciences AB.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Certifications/Compliance
ISO9001 and ISO13485
Concentration
30 mg/mL
Contents
Dynabeads Protein G are supplied in PBS, pH 7.4 w/0.01% Tween-20, 0.09% NaN3
Description
Recombinant Protein G covalently bound to Dynabeads
For Use With (Application)
Immunoprecipitation
Material
Polystyrene
Product Type
Superparamagnetic Beads
Format
Beads in suspension
For Use With (Equipment)
KingFisher™ Sample Purification System, DynaMag™ magnets
High-throughput Compatibility
High-throughput Compatible
Isolation Technology
Magnetic Beads
Ligand Type
Protein G
No. of Tests
20 Tests
Preservative
0.02% NaN3
Product Line
Dynabeads™
Purification Target
Antibodies
Purity or Quality Grade
For Research Use Only
Shelf Life
48 months from manufacturing date
Shipping Condition
Ambient temperature
Surface Functionality
Protein G
Uniformity
Monosized 2.8 mm (CV <5%)
Volume (Metric)
1 mL
Diameter (Metric)
2.8 μm

Contents & Storage

Store at 2–8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text