DRI™ pH-Detect™ Specimen Validity Tests
Product Image
Thermo Scientific™
DRI™ pH-Detect™ Specimen Validity Tests
Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ pH-Detect Specimen Validity Tests.
Have Questions?
Catalog number 100054
Price (USD)/ Each
-
Request A Quote
Description:
DRI pH-Detect Assay Specimen Validity Test
Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ pH-Detect Specimen Validity Tests. Liquid and ready-to-use, these tests can be performed on a variety of general chemistry analyzers.
The use of additives and adulterants to mask controlled substances in urine samples is a difficult and ongoing challenge to drugs of abuse screening. We offer the industry's most extensive menu of detection tests. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution. The adulteration assays provide a complete and automated way to test for sample integrity and adulteration.
  • Complete and automated way to test for sample integrity and adulteration
  • Associated calibrators and controls offered in convenient sizes
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionDRI pH-Detect Assay Specimen Validity Test
Detectable AnalytespH
Sample TypeUrine
TypeReagent
Quantity2 x 500 mL
Shelf LifeUntil Label Expiry Date
Unit SizeEach
Contents & Storage
2°C to 8°C