CEDIA™ Barbiturate Drugs of Abuse Assays
Product Image
Thermo Scientific™

CEDIA™ Barbiturate Drugs of Abuse Assays

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Barbiturate Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100084TS3 x 17 mL, TSC
1000931 x 65 mL, MCC
100173653 x 17 mL, Indiko
16612131 x 495 mL, LC
Catalog number 100084TS
Price (USD)
-
Request A Quote
Quantity:
3 x 17 mL, TSC
Ask our AI about this Product
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Barbiturate Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA™ assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • One of the largest menus of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionCEDIA Barbiturate Drugs of Abuse Assays
Detectable AnalytesBarbiturate
DoA CalibratorsCEDIA Multi-drug Calibrators
Sample TypeUrine
TypeReagent
Quantity3 x 17 mL, TSC
Unit SizeEach
Contents & Storage
2°C to 8°C
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
75381213_123127Certificate of AnalysisMar 24, 2025100093
75381212_123127Certificate of AnalysisMar 10, 202510017365
74765454Certificate of AnalysisJan 29, 2025100093
74984805Certificate of AnalysisJan 29, 202510017365
74984806Certificate of AnalysisJan 29, 2025100093
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection