Invitrogen™

Attune™ NxT Waste Bottle

Catalog number: 100085928CST
Have Questions?
Invitrogen™

Attune™ NxT Waste Bottle

Catalog number: 100085928CST
Have Questions?
Catalog Number
100085928CST
Unit Size
1 bottle
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
100085928CST1 bottle
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This is a 1.9-L replacement bottle for the waste tank on the Attune NxT Acoustic Focusing Cytometer. The bottle is designed for easy user replacement and with more robust fittings to prevent breakage. This bottle replaces discontinued Cat. No. 100022156.

Specifications

For Use With (Equipment)
Attune NxT
Includes
Waste Bottle
Product Line
FPL
Quantity
1 bottle
Shipping Condition
Room temperature
Product Type
Flow Cytometer Accessory

Contents & Storage

1 Bottle

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text