Invitrogen™

Attune™ Nxt Bottle Purge Filter

Catalog number: 100087959CST
Have Questions?
Invitrogen™

Attune™ Nxt Bottle Purge Filter

Catalog number: 100087959CST
Have Questions?
Catalog Number
100087959CST
Unit Size
1 filter
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
100087959CST1 filter
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This is a replacement bottle purge filter for use on the Attune NxT Acoustic Focusing Cytometer. The purge filter allows air into the bottle to prevent back pressure buildup. The filter limits escape of aerosols from the waste bottle and can be replaced easily if contaminated.

Specifications

For Use With (Equipment)
Attune NxT
Format
Muffler
Includes
1 Muffler
Product Line
FPL
Quantity
1 filter
Shipping Condition
Room temperature
Product Type
Flow Cytometer Accessory

Contents & Storage

1 Filter

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text