Invitrogen™

Attune™ NxT Level Sensor Cable

Catalog number: 100087973CST
Have Questions?
Invitrogen™

Attune™ NxT Level Sensor Cable

Catalog number: 100087973CST
Have Questions?
Catalog Number
100087973CST
Unit Size
1 cable
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
100087973CST1 cable
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This is a replacement level sensor cable for the focusing fluid and waste bottles of the Attune NxT Acoustic Focusing Cytometer. It can be replaced easily without replacing the entire bottle assembly if the sensor connector is broken.

Specifications

For Use With (Equipment)
Attune NxT
Format
Sensor cable
Product Line
FPL
Shipping Condition
Room Temperature
Product Type
Flow Cytometer Accessory

Contents & Storage

1 Cable

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text