PBS, pH 7.4
PBS, pH 7.4
Gibco™

PBS, pH 7.4

Gibco PBS, pH 7.4, is a balanced salt solution ideal for cell culture applications. It lacks calcium, magnesium, and phenol red, preventing cell adhesion and assay interference. Trusted for quality, Gibco PBS ensures reliable results in cell washing, transport, and reagent preparation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
10010023500 mL
100100311000 mL
1001004910 x 500 mL
100100726 x 1000 mL
Catalog number 10010023
Price (USD)
30.65
Online Exclusive
34.10
Save 3.45 (10%)
Each
Add to cart
Quantity:
500 mL
Price (USD)
30.65
Online Exclusive
34.10
Save 3.45 (10%)
Each
Add to cart

Gibco Phosphate Buffered Saline (PBS), pH 7.4, is a balanced salt solution widely used in various cell culture applications. This product is specifically formulated without calcium, magnesium, and phenol red, making it ideal for a range of laboratory procedures.

Key features

  • Without calcium and magnesium—the absence of calcium and magnesium prevents unwanted cell adhesion and aggregation, which is crucial during cell dissociation and flow cytometry
  • Without phenol red—the lack of phenol red ensures that there is no interference with optical measurements and prevents potential effects on cell physiology

Applications

  • Washing cells before dissociation—PBS is commonly used to rinse cells before enzymatic dissociation, ensuring that chelators are removed from the culture
  • Transporting cells or tissue—PBS provides a stable environment for cells or tissues during transportation
  • Diluting cells for counting—PBS is used to dilute cell suspensions for accurate counting
  • Preparing reagents—PBS serves as a buffer in the preparation of various reagents

Why choose PBS over DPBS

  • Simplicity—PBS is a simpler formulation compared to Dulbecco's Phosphate-Buffered Saline (DPBS), which includes additional components like potassium chloride and may be formulated with or without calcium, magnesium, glucose, and pyruvate. This simplicity makes PBS a versatile choice for general cell culture applications.
  • Specific applications—PBS is preferred for applications where the presence of potassium or other additives in DPBS might interfere with experimental outcomes

Why choose pH 7.4 over pH 7.2

  • Physiological relevance—pH 7.4 is closer to the physiological pH of human blood, making it more suitable for maintaining the natural state of cells during experiments. This pH is particularly important for applications involving mammalian cells.

Why choose Gibco PBS, pH 7.4

  • Quality and reliability—manufacture of Gibco PBS, pH 7.4, is cGMP-compliant under the ISO 13485 standard, ensuring high standards of quality and consistency
  • Extensive testing—lot release testing of osmolality, pH levels, stability, endotoxins, and sterility ensures the quality of the product
  • Proven track record—Gibco cell culture products have a long-standing reputation in the scientific community, with numerous publications citing the use of Gibco PBS, reflecting its widespread acceptance and trust among researchers

Gibco PBS, pH 7.4, is a reliable and high-quality solution for various applications. Its formulation without calcium, magnesium, and phenol red makes it versatile and suitable for specific experimental needs. Choosing Gibco PBS ensures consistency, quality, and support from a trusted brand in the scientific community.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialPhosphate Buffered Saline (PBS)
ColorClear
IncludesNo Calcium, No Magnesium
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality280 - 315 mOsm/kg
PackagingBottle
Recommended StorageStorage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
SterilitySterile-filtered
Concentration1 X
For Use With (Application)Washing Cells Before Dissociation; Diluting Cells for Counting and Preparing Reagents; Transporting Cells or Tissue
Physical FormLiquid
Product LineGibco
Quantity500 mL
Solution TypePhosphate Buffered Saline
pH7.4
Unit SizeEach

Frequently asked questions (FAQs)

How can I make isolation buffer for cell isolation?

To prepare this buffer:

(1) Make up 100 mL of a stock solution of 0.5 M EDTA by dissolving 14.6 g EDTA in approximately 80 mL of ddH2O. Adjust the pH to 7.4, then adjust the volume to 100 mL. Autoclave and filter 25 mL aliquots through a sterilized filter into 4 sterile 50 mL tubes. Store one at 4 degrees C and the rest at -20 degrees C for future use.
(2) Make up a stock solution of 20% BSA by putting 5 g of BSA in a 50 mL tube and filling it with ddH2O up to 25 mL. Leave on a roller at room temperature until dissolved. Filter through a sterile 0.2 µm filter into a sterile 50 mL tube and adjust the volume to 30 mL with sterile H2O.
(3) Make the isolation buffer by taking a 500 mL bottle of 1x PBS, pH 7.4 without Ca2+ or Mg2+ (Cat. No. 10010023). Add 2.5 mL 20% BSA and 2 mL 0.5 M EDTA. Mix by turning the bottle end over end 10 times. Store at 4 degrees C.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

My PBS, PH 7.4 (Cat. No. 10010072) was accidentally frozen. Can I still use it?

This product should be stored at 2-8 degrees C and should not be frozen. The biggest issue with accidentally freezing this product is its solubility. If this product was accidentally frozen, we recommend placing it in a 2-8 degree environment and allowing it to slowly thaw overnight. If the product is fully thawed and shows no signs of precipitation, then it should still be usable, but we cannot guarantee effectiveness.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your PBS products tested for endotoxin levels?

Our PBS, pH 7.2 and PBS, pH 7.4 products are tested for endotoxin and the endotoxin levels are shown on the Certificate of Analysis (CoA) for the particular lot number of the product.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your PBS products sterile and suitable for cell culture?

We offer PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free (Cat. Nos. AM9624 and AM9625), which is molecular biology grade and is certified to be RNase-free. However, it is not sterile and hence is not cell culture-grade.

All our other PBS products (Cat. Nos. 70011xxx, 10010xxx, 70013xxx, 20012xxx) are filter-sterilized using a 0.1 µm filter and are cell culture grade (ready to use directly on cells). However, they are not tested for nuclease activity.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your PBS products DNase- and RNase-free?

We offer PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free (Cat. Nos. AM9624 and AM9625) that is molecular biology grade and is certified to be RNase-free. Our other PBS products are made with distilled water and are filter-sterilized, but are not tested for RNase and DNase activity.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (6)

Citations & References
Abstract
A comprehensive protocol for efficient differentiation of human NPCs into electrically competent neurons.
Authors:Romito E,Battistella I,Plakhova V,Paplekaj A,Forastieri C,Toffolo E,Musio C,Conti L,Battaglioli E,Rusconi F
Journal:Journal of neuroscience methods
PubMed ID:39053772
Aldose reductase mediates mitogenic signaling in vascular smooth muscle cells.
Authors: Ramana Kota V; Chandra Deepak; Srivastava Sanjay; Bhatnagar Aruni; Aggarwal Bharat B; Srivastava Satish K;
Journal:J Biol Chem
PubMed ID:12063254
'Abnormal vascular smooth muscle cell (VSMC) proliferation is a key feature of atherosclerosis and restenosis; however, the mechanisms regulating growth remain unclear. Herein we show that inhibition of the aldehyde-metabolizing enzyme aldose reductase (AR) inhibits NF-kappa B activation during restenosis of balloon-injured rat carotid arteries as well as VSMC proliferation ... More
Blocking HES1 expression initiates GABAergic differentiation and induces the expression of p21(CIP1/WAF1) in human neural stem cells.
Authors: Kabos Peter; Kabosova Andrea; Neuman Toomas;
Journal:J Biol Chem
PubMed ID:11809764
Mammalian neural stem cells can develop into a variety of neuronal and glial cell types. This involves a highly coordinated process of differentiation in which the Notch signaling pathway and the system of helix-loop-helix (HLH) transcriptional regulators play a key role. By exercising control over proliferation, initiation of differentiation, neurite ... More
Cholesterol modulates the membrane binding and intracellular distribution of annexin 6.
Authors: de Diego Iñaki; Schwartz Felix; Siegfried Heide; Dauterstedt Paul; Heeren Joerg; Beisiegel Ulrike; Enrich Carlos; Grewal Thomas;
Journal:J Biol Chem
PubMed ID:12070178
Annexins are Ca(2+)- and phospholipid-binding proteins that are widely expressed in mammalian tissues and that bind to different cellular membranes. In recent years its role in membrane traffic has emerged as one of its predominant functions, but the regulation of its intracellular distribution still remains unclear. We demonstrated that annexin ... More
A 2-D liquid separations/mass mapping method for interlysate comparison of ovarian cancers.
Authors: Kachman Maureen T; Wang Haixing; Schwartz Donald R; Cho Kathleen R; Lubman David M;
Journal:Anal Chem
PubMed ID:11985308
A two-dimensional liquid phase separation of proteins from whole cell lysates coupled on-line to an electrospray-ionization time-of-flight (ESI-TOF) mass spectrometer (MS) is used to map the protein content of ovarian surface epithelial cells (OSE) and an ovarian carcinoma-derived cell line (ES2). The two dimensions involve the use of liquid isoelectric ... More