Search Thermo Fisher Scientific
Compatibility | Use this part either as a spare/replacement part or if you already have a Smart Rotary Cell Changer Base (e.g. included with a CVC) |
Description | Spare 7-cell Changer, 7-position rotary turret |
For Use With (Equipment) | Smart Rotary Cell Changer Base |
Unit Size | Each |
Compatibility | Compatible with temperature probe accessory for Smart accessories, catalog#10011401 |
Configuration Details | Evolution One/One Plus- sample position only, Evolution Pro- sample and reference positions |
Description | Smart Thermostatted Rotary 7-Cell Changer |
Type | Smart Thermostatted 7-Cell Rotary Changer |
For Use With (Equipment) | Evolution One/One Plus Series and Evolution Pro spectrophotometers |
Includes | Carousel and motor base |
Thermostatting | Thermostatting with flowing liquid from an external temperature controlled recirculator |
Unit Size | Each |
Configuration Details | Single temperature probe. Connects directly to the Smart accessory motor base. Monitors the temperature in one cuvette. |
Description | Temperature probe accessory for Smart accessories |
Type | Temperature Probe |
For Use With (Equipment) | Smart Rotary Cell Changer Base |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
10010110 | Each | Spare 7-cell Changer, 7-position rotary turret | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
840-214900 | Each | Smart Thermostatted Rotary 7-Cell Changer | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
10011401 | Each | Temperature probe accessory for Smart accessories | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|