Search Thermo Fisher Scientific
Description | Fiber Optic Coupler for Evolution Pro (only) Passive fiber optic coupler – uses a mirror and a lenses to couple light from the monochromator to the input fiber of the fiber probe. A second lens and mirror directs returned light to the instruments standard detector. |
For Use With (Equipment) | Thermo Scientific Evolution Pro Spectrophotometer |
Unit Size | Each |
Description | Integrated Fiber Optics Module for Evolution One or Evolution One Plus Integrated Fiber Optics Module is designed to deliver maximum performance from any fiber optic probe. A single lens, matched to the Fiber AFBG setting, couples light from the monochromator to the input fiber of the fiber probe. Light from the returned fiber shines directly onto a detector built into the module. Eliminating optical components while maximizing energy throughput delivers performance beyond expectations for a fiber probe system. |
For Use With (Equipment) | Evolution One or Evolution One Plus Spectrophotometers (only) |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | For Use With (Equipment) | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
10012201 | Each | Fiber Optic Coupler for Evolution Pro (only) Passive fiber optic coupler – uses a mirror and a lenses to couple light from the monochromator to the input fiber of the fiber probe. A second lens and mirror directs returned light to the instruments standard detector. | Thermo Scientific Evolution Pro Spectrophotometer | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
840-211400 | Each | Integrated Fiber Optics Module for Evolution One or Evolution One Plus Integrated Fiber Optics Module is designed to deliver maximum performance from any fiber optic probe. A single lens, matched to the Fiber AFBG setting, couples light from the monochromator to the input fiber of the fiber probe. Light from the returned fiber shines directly onto a detector built into the module. Eliminating optical components while maximizing energy throughput delivers performance beyond expectations for a fiber probe system. | Evolution One or Evolution One Plus Spectrophotometers (only) | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Fiber Optic Probes allow you to measure your sample in its container without requiring transfer of the sample to a cuvette and insertion of the cuvette into the sample compartment of the spectrophotometer.
Advantages of the Evolution One/One Plus Integrated Fiber Optics Module
Advantages of fiber optic probe measurements
Limitations when working with Fiber Optic Probes